Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011672.2 | NP_001011672.2 | 1 - 2 | 115 | 100 | |
GenBank mRNA | BC113355.1 | - | 1 - 2 | 115 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |