Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011676.3 | NP_001011676.2 | 1 - 2 | 91 | 67 | |
GenBank mRNA | AY841149.1 | - | 1 - 2 | 88 | 67 | |
BC118117.1 | - | 1 - 2 | 92 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |