Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011676.3 | NP_001011676.2 | 2 - 3 | 394 | 150 | |
GenBank mRNA | AY841149.1 | - | 2 - 3 | 391 | 150 | |
BC118117.1 | - | 2 - 3 | 395 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |