Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014878.2 | NP_001014878.2 | 1 - 2 | 190 | 74 | |
GenBank mRNA | BC114006.1 | - | 1 - 2 | 190 | 74 | |
BT020944.1 | - | 1 - 2 | 185 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |