Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015627.2 | NP_001015627.1 | 6 - 7 | 1163 | 110 | |
GenBank mRNA | BC118313.1 | - | 6 - 7 | 1163 | 110 | |
BT020922.1 | - | 6 - 7 | 1121 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |