Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034232.2 | NP_001029404.1 | 6 - 7 | 600 | 139 | |
GenBank mRNA | BC113322.1 | - | 6 - 7 | 600 | 139 | |
BT021574.1 | - | 6 - 7 | 568 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |