Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035328.2 | NP_001030405.2 | 1 - 2 | 93 | 68 | |
GenBank mRNA | BC113297.1 | - | 1 - 2 | 93 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |