Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045920.1 | NP_001039385.1 | 4 - 5 | 261 | 133 | |
GenBank mRNA | BC113259.1 | - | 4 - 5 | 261 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |