Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045930.2 | NP_001039395.1 | 5 - 6 | 759 | 91 | |
GenBank mRNA | BC114057.1 | - | 5 - 6 | 760 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |