Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045941.1 | NP_001039406.1 | 3 - 4 | 789 | 77 | |
GenBank mRNA | BC105455.1 | - | 3 - 4 | 789 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |