Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045983.1 | NP_001039448.1 | 1 - 2 | 140 | 60 | |
GenBank mRNA | BC113230.1 | - | 1 - 2 | 140 | 60 | |
KT351731.1 | - | 1 - 2 | 140 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |