Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045992.2 | NP_001039457.2 | 2 - 3 | 310 | 109 | |
NR_028054.1 | 1 - 2 | 693 | 109 | |||
GenBank mRNA | BC113206.1 | - | 1 - 2 | 211 | 109 | |
DT816311.1 | - | 1 - 2 | 693 | 109 | ||
DT856026.1 | - | 2 - 3 | 311 | 109 | ||
EV660199.1 | - | 2 - 3 | 292 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |