Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045996.2 | NP_001039461.1 | 5 - 6 | 692 | 71 | |
GenBank mRNA | BC105311.1 | - | 5 - 6 | 693 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |