Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046001.2 | NP_001039466.1 | 2 - 3 | 621 | 90 | |
GenBank mRNA | BC114085.1 | - | 2 - 3 | 622 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |