Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046008.2 | NP_001039473.1 | 1 - 2 | 139 | 66 | |
GenBank mRNA | BC112465.1 | - | 1 - 2 | 141 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |