Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046020.2 | NP_001039485.1 | 4 - 5 | 663 | 143 | |
GenBank mRNA | BC114047.1 | - | 4 - 5 | 666 | 143 | |
FJ594406.1 | - | 3 - 4 | 373 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |