Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046045.1 | NP_001039510.1 | 3 - 4 | 313 | 76 | |
GenBank mRNA | BC113336.1 | - | 3 - 4 | 313 | 76 | |
BT026267.1 | - | 3 - 4 | 390 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |