Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046068.2 | NP_001039533.1 | 4 - 5 | 502 | 107 | |
GenBank mRNA | BC105368.1 | - | 4 - 5 | 503 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |