Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046073.2 | NP_001039538.1 | 11 - 12 | 1652 | 143 | |
GenBank mRNA | BC114127.1 | - | 11 - 12 | 1653 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |