Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046118.2 | NP_001039583.1 | 7 - 8 | 779 | 65 | |
GenBank mRNA | BC105438.1 | - | 7 - 8 | 781 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |