Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046130.2 | NP_001039595.1 | 6 - 7 | 822 | 99 | |
GenBank mRNA | BC105444.1 | - | 6 - 7 | 823 | 99 | |
BT025352.1 | - | 6 - 7 | 705 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |