Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046138.2 | NP_001039603.1 | 2 - 3 | 210 | 80 | |
GenBank mRNA | BC114859.1 | - | 2 - 3 | 211 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |