Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046147.1 | NP_001039612.1 | 7 - 8 | 656 | 136 | |
GenBank mRNA | BC113324.1 | - | 7 - 8 | 656 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |