Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046148.1 | NP_001039613.1 | 11 - 12 | 1109 | 114 | |
GenBank mRNA | BC112777.1 | - | 11 - 12 | 1109 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |