Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046169.1 | NP_001039634.1 | 1 - 2 | 51 | 65 | |
GenBank mRNA | BC113350.1 | - | 1 - 2 | 51 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |