Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046209.1 | NP_001039674.1 | 6 - 7 | 692 | 76 | |
GenBank mRNA | BC114066.1 | - | 6 - 7 | 692 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |