Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046224.1 | NP_001039689.1 | 1 - 2 | 220 | 119 | |
GenBank mRNA | BC114087.1 | - | 1 - 2 | 220 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |