Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046228.1 | NP_001039693.1 | 1 - 2 | 137 | 80 | |
GenBank mRNA | BC112781.1 | - | 1 - 2 | 137 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |