Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046267.1 | NP_001039732.1 | 2 - 3 | 422 | 60 | |
GenBank mRNA | BC112503.1 | - | 2 - 3 | 422 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |