Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046296.2 | NP_001039761.1 | 7 - 8 | 1560 | 72 | |
GenBank mRNA | BC114148.1 | - | 7 - 8 | 1561 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |