Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046299.1 | NP_001039764.1 | 3 - 4 | 559 | 79 | |
GenBank mRNA | DQ421787.1 | - | 3 - 4 | 559 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |