Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C29H11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046315.2 | NP_001039780.1 | 2 - 3 | 96 | 77 | |
GenBank mRNA | BC105390.1 | - | 2 - 3 | 97 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |