Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046317.2 | NP_001039782.1 | 6 - 7 | 1357 | 106 | |
GenBank mRNA | BC112826.1 | - | 6 - 7 | 1358 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |