Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046336.1 | NP_001039801.1 | 5 - 6 | 667 | 58 | |
GenBank mRNA | BC114078.1 | - | 5 - 6 | 667 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |