Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046378.1 | NP_001039843.1 | 2 - 3 | 376 | 136 | |
GenBank mRNA | BC112806.1 | - | 2 - 3 | 376 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |