Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046379.1 | NP_001039844.1 | 2 - 3 | 244 | 69 | |
GenBank mRNA | BC113327.1 | - | 2 - 3 | 244 | 69 | |
BT030714.1 | - | 2 - 3 | 238 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |