Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046410.1 | NP_001039875.1 | 3 - 4 | 425 | 92 | |
GenBank mRNA | BC113253.1 | - | 3 - 4 | 425 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |