Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046411.1 | NP_001039876.1 | 8 - 9 | 1272 | 64 | |
GenBank mRNA | BC113214.1 | - | 8 - 9 | 1272 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |