Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046449.1 | NP_001039914.1 | 5 - 6 | 933 | 83 | |
GenBank mRNA | BC105400.1 | - | 5 - 6 | 933 | 83 | |
BT029904.1 | - | 4 - 5 | 755 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |