Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046515.2 | NP_001039980.1 | 1 - 2 | 196 | 71 | |
GenBank mRNA | BC114793.1 | - | 1 - 2 | 197 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |