Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046530.2 | NP_001039995.2 | 4 - 5 | 438 | 121 | |
GenBank mRNA | BC114134.1 | - | 4 - 5 | 439 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |