Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046538.1 | NP_001040003.1 | 4 - 5 | 474 | 83 | |
GenBank mRNA | BC114760.1 | - | 4 - 5 | 474 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |