Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046557.2 | NP_001040022.1 | 2 - 3 | 225 | 66 | |
GenBank mRNA | BC112500.1 | - | 2 - 3 | 226 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |