Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOTA-T2R10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046627.1 | NP_001040092.1 | 1 | 456 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |