Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075130.1 | NP_001068598.1 | 3 - 4 | 520 | 65 | |
GenBank mRNA | AF135233.1 | - | 2 - 3 | 224 | 65 | |
BC118075.1 | - | 3 - 4 | 520 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |