Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075139.1 | NP_001068607.1 | 7 - 8 | 579 | 87 | |
GenBank mRNA | AM075206.1 | - | 7 - 8 | 579 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |