Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075163.1 | NP_001068631.1 | 6 - 7 | 515 | 89 | |
GenBank mRNA | BC118224.1 | - | 6 - 7 | 515 | 89 | |
BC151415.1 | - | 6 - 7 | 536 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |