Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075173.1 | NP_001068641.1 | 5 - 6 | 685 | 76 | |
GenBank mRNA | BC112659.1 | - | 5 - 6 | 685 | 76 | |
DQ087595.1 | - | 1 - 2 | 146 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |