Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075173.1 | NP_001068641.1 | 8 - 9 | 1161 | 104 | |
GenBank mRNA | BC112659.1 | - | 8 - 9 | 1161 | 104 | |
DQ087595.1 | - | 4 - 5 | 622 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |