Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075251.1 | NP_001068719.1 | 3 - 4 | 279 | 122 | |
GenBank mRNA | BC118233.1 | - | 3 - 4 | 279 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |